Assay Details
Target Gene Details
Entrez Gene ID: | 55024 |
Gene Name: | B-cell scaffold protein with ankyrin repeats 1 |
Gene Aliases: |
BANK |
Location: |
Chr.4:101790482-102074812 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BANK1 | NM_001127507.2 | NP_001120979.2 | ||
| NM_017935.4 | NP_060405.4 | |||
| XM_017008337.1 | XP_016863826.1 | |||
| AK091523.1 | BAC03685.1 | |||
| AK292133.1 | ||||
| AL834291.1 | CAD38965.2 | |||
| BC032241.1 | AAH32241.2 | |||
| DA109829.1 | ||||
| EU051376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759269 | Chr.4:101741542 - 102087783 on Build GRCh38 | Gain |
|
| nsv999861 | Chr.4:101654958 - 102009405 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map