Assay Details
Target Gene Details
Entrez Gene ID: | 5783 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 13 |
Gene Aliases: |
FAP-1, PNP1, PTP-BAS, PTP-BL, PTP1E, PTPL1, PTPLE, hPTP1E |
Location: |
Chr.4:86594307-86815179 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN13 | NM_006264.2 | NP_006255.1 | ||
| NM_080683.2 | NP_542414.1 | |||
| NM_080684.2 | NP_542415.1 | |||
| NM_080685.2 | NP_542416.1 | |||
| XM_011532165.1 | XP_011530467.1 | |||
| XM_017008511.1 | XP_016864000.1 | |||
| XM_017008512.1 | XP_016864001.1 | |||
| XM_017008513.1 | XP_016864002.1 | |||
| XM_017008514.1 | XP_016864003.1 | |||
| XM_017008515.1 | XP_016864004.1 | |||
| XM_017008516.1 | XP_016864005.1 | |||
| AK056151.1 | ||||
| D21209.1 | BAA04750.1 | |||
| D21210.1 | BAA04751.1 | |||
| D21211.1 | BAA04752.1 | |||
| DA625835.1 | ||||
| X80289.1 | CAA56563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759264 | Chr.4:86496138 - 86768380 on Build GRCh38 | Loss |
|
| nsv594786 | Chr.4:86155207 - 86688167 on Build GRCh38 | Gain |
|
| esv3601257 | Chr.4:86557520 - 86614379 on Build GRCh38 | Loss |
|
| esv3601258 | Chr.4:86558468 - 86701058 on Build GRCh38 | Loss |
|
| nsv822635 | Chr.4:86558423 - 86701023 on Build GRCh38 | Loss |
|
| nsv594787 | Chr.4:86300674 - 86967515 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80184743] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map