Assay Details
Target Gene Details
Entrez Gene ID: | 643783 |
Gene Name: | USP46 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:52659406-52661498 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP46-AS1 | NR_125363.1 | |||
| AA405370.1 | ||||
| BX354350.2 | ||||
| CN313378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524494 | Chr.4:52593454 - 52715898 on Build GRCh38 | Loss |
|
| nsv594201 | Chr.4:51932458 - 52866278 on Build GRCh38 | Loss |
|

Genomic Map