Assay Details
Target Gene Details
Entrez Gene ID: | 57733 |
Gene Name: | glucosylceramidase beta 3 (gene/pseudogene) |
Gene Aliases: |
CBG, CBGL1, GLUC, KLRP |
Location: |
Chr.4:22692914-22819575 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBA3 | NM_001128432.2 | NP_001121904.1 | ||
| NM_020973.4 | NP_066024.1 | |||
| NR_102355.1 | ||||
| NR_102356.1 | ||||
| AB017913.1 | BAB18741.1 | |||
| AF317840.1 | AAG39217.1 | |||
| AF323990.1 | AAL37305.1 | |||
| AJ278964.1 | CAC08178.1 | |||
| AK222963.1 | BAD96683.1 | |||
| AK292746.1 | ||||
| AK313189.1 | ||||
| BC029362.1 | AAH29362.1 | |||
| BC070188.1 | AAH70188.1 | |||
| BC101829.1 | AAI01830.1 | |||
| BC109377.1 | AAI09378.1 | |||
| BC136264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2664670 | Chr.4:22699126 - 22775543 on Build GRCh38 | Deletion |
|
| nsv821634 | Chr.4:22425451 - 22864325 on Build GRCh38 | Loss |
|
| nsv829883 | Chr.4:22664703 - 22840894 on Build GRCh38 | Loss |
|
| esv3599912 | Chr.4:22699143 - 22775511 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73109790,rs74715385] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map