Assay Details
Target Gene Details
Entrez Gene ID: | 79804 |
Gene Name: | HAND2 antisense RNA 1 (head to head) |
Gene Aliases: |
DEIN, NBLA00301 |
Location: |
Chr.4:173528600-173591255 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAND2-AS1 | NR_003679.2 | |||
| NR_136192.1 | 4 | 2225 | ||
| NR_136193.1 | 4 | 2922 | ||
| NR_136194.1 | 4 | 2392 | ||
| NR_136195.1 | ||||
| NR_136196.1 | ||||
| NR_136197.1 | 4 | 2296 | ||
| NR_136198.1 | 3 | 2113 | ||
| NR_136199.1 | 5 | 2435 | ||
| NR_136200.1 | 4 | 2925 | ||
| NR_136201.1 | 4 | 3092 | ||
| AK054605.1 | ||||
| AK123668.1 | ||||
| DA863153.1 | ||||
| DB292439.1 | ||||
| DQ831955.1 | 1 | 985 | ||
| DQ831956.1 | 1 | 985 | ||
| EF529495.1 | 4 | 1225 | ||
| EF529496.1 | 4 | 1148 | ||
| EF529497.1 | 4 | 1096 | ||
| EF529498.1 | 4 | 1360 | ||
| EF529499.1 | ||||
| EF529500.1 | ||||
| EF529501.1 | ||||
| EF529502.1 | ||||
| EF529503.1 | ||||
| EF529504.1 | ||||
| EF529505.1 | ||||
| EU000853.1 | 4 | 2138 | ||
| EU000854.1 | 4 | 2138 | ||
| EU000855.1 | 3 | 3230 | ||
| EU000856.1 | 3 | 3230 |

Genomic Map