Assay Details
Target Gene Details
Entrez Gene ID: | 5311 |
Gene Name: | polycystin 2, transient receptor potential cation channel |
Gene Aliases: |
APKD2, PC2, PKD4, Pc-2, TRPP2 |
Location: |
Chr.4:88007647-88077779 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD2 | NM_000297.3 | NP_000288.1 | ||
| XM_011532028.2 | XP_011530330.1 | |||
| XM_011532029.1 | XP_011530331.1 | |||
| BC112261.1 | AAI12262.1 | |||
| BC112263.1 | AAI12264.1 | |||
| U50928.1 | AAC50520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2665331 | Chr.4:87937174 - 88153583 on Build GRCh38 | Deletion |
|
| esv2759266 | Chr.4:87997399 - 88291967 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115145783] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map