Assay Details
Target Gene Details
Entrez Gene ID: | 497189 |
Gene Name: | TIFA inhibitor |
Gene Aliases: |
- |
Location: |
Chr.5:135448868-135452399 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIFAB | NM_001099221.1 | NP_001092691.1 | ||
| AB161513.1 | BAD89018.1 |
More Information
Set Membership: |
|

Genomic Map