Assay Details
Target Gene Details
Entrez Gene ID: | 441094 |
Gene Name: | NR2F1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:93409356-93581320 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR2F1-AS1 | NR_021491.2 | |||
| NR_109820.1 | ||||
| NR_109821.1 | ||||
| NR_109822.1 | ||||
| NR_109823.1 | ||||
| NR_109824.1 | ||||
| NR_109825.1 | ||||
| AK124699.1 | ||||
| AK296281.1 | ||||
| AK311143.1 | ||||
| BX647510.1 | ||||
| DB257894.1 | ||||
| HY108556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830402 | Chr.5:93368195 - 93511639 on Build GRCh38 | Gain |
|

Genomic Map