Assay Details
Target Gene Details
Entrez Gene ID: | 1719 |
Gene Name: | dihydrofolate reductase |
Gene Aliases: |
DHFRP1, DYR |
Location: |
Chr.5:80626226-80654981 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHFR | NM_000791.3 | NP_000782.1 | ||
NM_001290354.1 | NP_001277283.1 | |||
NM_001290357.1 | NP_001277286.1 | |||
NR_110936.1 | ||||
AK293146.1 | ||||
AK297302.1 | ||||
AK308492.1 | ||||
BC000192.1 | AAH00192.1 | |||
BC003584.1 | AAH03584.2 | |||
BC070280.1 | AAH70280.1 | |||
BC071996.1 | AAH71996.1 | |||
J00140.1 | AAA58485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483112 | Chr.5:80551019 - 80731514 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv4895 | Chr.5:80623591 - 80640149 on Build GRCh38 | Insertion |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
