Assay Details
Target Gene Details
Entrez Gene ID: | 2264 |
Gene Name: | fibroblast growth factor receptor 4 |
Gene Aliases: |
CD334, JTK2, TKF |
Location: |
Chr.5:177086872-177098144 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGFR4 | NM_001291980.1 | NP_001278909.1 | ||
| NM_002011.4 | NP_002002.3 | |||
| NM_213647.2 | NP_998812.1 | |||
| XM_005265838.3 | XP_005265895.1 | |||
| XM_011534464.2 | XP_011532766.1 | |||
| XM_011534465.2 | XP_011532767.1 | |||
| AB209631.1 | 1 | 538 | BAD92868.1 | |
| AK225280.1 | ||||
| AK225909.1 | ||||
| AK301169.1 | ||||
| BC011847.2 | AAH11847.1 | |||
| BM453707.1 | ||||
| L03840.1 | AAB59389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823351 | Chr.5:177079996 - 177144595 on Build GRCh38 | Gain |
|
| dgv363n21 | Chr.5:176931607 - 177242486 on Build GRCh38 | Gain |
|
| nsv509101 | Chr.5:177044099 - 177135363 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map