Assay Details
Target Gene Details
Entrez Gene ID: | 57472 |
Gene Name: | CCR4-NOT transcription complex subunit 6 |
Gene Aliases: |
CCR4, Ccr4a |
Location: |
Chr.5:180494373-180578405 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT6 | NM_001303241.1 | NP_001290170.1 | ||
| XM_005265953.1 | XP_005266010.1 | |||
| XM_011534605.2 | XP_011532907.1 | |||
| XM_011534606.1 | XP_011532908.1 | |||
| XM_011534607.1 | XP_011532909.1 | |||
| XM_017009672.1 | XP_016865161.1 | |||
| AB033020.1 | ||||
| BC152469.1 | ||||
| BQ232180.1 | ||||
| DA370007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019519 | Chr.5:180439730 - 180532408 on Build GRCh38 | Gain |
|
| nsv830564 | Chr.5:180440394 - 180583096 on Build GRCh38 | Gain |
|
| nsv509110 | Chr.5:180425156 - 180510675 on Build GRCh38 | Insertion |
|
| nsv1026417 | Chr.5:180294376 - 180864360 on Build GRCh38 | Gain |
|
| esv3607782 | Chr.5:180443803 - 180540294 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114746674] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map