Assay Details
Target Gene Details
Entrez Gene ID: | 345757 |
Gene Name: | family with sequence similarity 174 member A |
Gene Aliases: |
HGS_RE408, TMEM157, UNQ1912 |
Location: |
Chr.5:100535305-100586741 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM174A | NM_198507.2 | NP_940909.1 | ||
| XM_006714600.1 | XP_006714663.1 | |||
| AK289539.1 | ||||
| AY359108.1 | AAQ89466.1 | |||
| BC027332.1 | AAH27332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763473 | Chr.5:100547648 - 100571960 on Build GRCh38 | Loss |
|
| nsv818363 | Chr.5:100563147 - 100570518 on Build GRCh38 | Gain |
|
| esv3890678 | Chr.5:100324445 - 101099725 on Build GRCh38 | Gain |
|
| nsv599165 | Chr.5:100133300 - 100727335 on Build GRCh38 | Loss |
|
| nsv1021183 | Chr.5:100290153 - 100885722 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76249414] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map