Assay Details
Target Gene Details
Entrez Gene ID: | 84105 |
Gene Name: | pterin-4 alpha-carbinolamine dehydratase 2 |
Gene Aliases: |
DCOH2, DCOHM, PHS2 |
Location: |
Chr.5:134905120-134962646 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBD2 | NM_032151.4 | NP_115527.3 | ||
| AF499009.1 | AAM18136.1 | |||
| AK098495.1 | ||||
| AL136721.1 | CAB66655.1 | |||
| BC054021.1 | AAH54021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830494 | Chr.5:134825197 - 134984298 on Build GRCh38 | Gain+Loss |
|
| nsv830493 | Chr.5:134667777 - 134926221 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map