Assay Details
Target Gene Details
Entrez Gene ID: | 497189 |
Gene Name: | TIFA inhibitor |
Gene Aliases: |
- |
Location: |
Chr.5:135448868-135452399 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIFAB | NM_001099221.1 | 2 | 919 | NP_001092691.1 |
| BX119085.1 | 1 | 367 |
More Information
Additional Information:
For this assay, SNP(s) [rs72787109] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map