Assay Details
Target Gene Details
Entrez Gene ID: | 57491 |
Gene Name: | aryl-hydrocarbon receptor repressor |
Gene Aliases: |
AHH, AHHR, bHLHe77 |
Location: |
Chr.5:304176-438291 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHRR | NM_001242412.1 | NP_001229341.1 | ||
| NM_020731.4 | NP_065782.2 | |||
| AB033060.1 | ||||
| BC151852.1 | ||||
| BC152406.1 | ||||
| EU293605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759315 | Chr.5:17263 - 595840 on Build GRCh38 | Loss |
|
| esv3894107 | Chr.5:81322 - 801057 on Build GRCh38 | Gain |
|
| nsv428458 | Chr.5:93781 - 338972 on Build GRCh38 | Loss |
|
| esv2752083 | Chr.5:28951 - 346521 on Build GRCh38 | Gain |
|
| nsv512845 | Chr.5:318792 - 318949 on Build GRCh38 | Insertion |
|
| nsv1030767 | Chr.5:154674 - 457840 on Build GRCh38 | Gain |
|
| nsv596614 | Chr.5:299506 - 438449 on Build GRCh38 | Gain |
|
| nsv509041 | Chr.5:314395 - 438288 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148497960] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map