Assay Details
Target Gene Details
Entrez Gene ID: | 821 |
Gene Name: | calnexin |
Gene Aliases: |
CNX, IP90, P90 |
Location: |
Chr.5:179678631-179731641 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CANX | XM_011534665.2 | XP_011532967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830560 | Chr.5:179612211 - 179748757 on Build GRCh38 | Loss |
|
| nsv5160 | Chr.5:179675417 - 179709494 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map