Assay Details
Target Gene Details
Entrez Gene ID: | 102546294 |
Gene Name: | uncharacterized LOC102546294 |
Gene Aliases: |
- |
Location: |
Chr.5:148268307-148383850 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102546294 | NR_105057.1 | |||
| AK054753.1 | ||||
| DA523579.1 |
Target Gene Details
Entrez Gene ID: | 153218 |
Gene Name: | serine peptidase inhibitor, Kazal type 13 (putative) |
Gene Aliases: |
HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3 |
Location: |
Chr.5:148268860-148286210 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK13 | NM_001040129.2 | NP_001035218.1 | ||
| AK001520.1 | ||||
| BC128163.1 | ||||
| BC128164.1 | ||||
| DQ438947.1 | ABD96086.1 | |||
| EF108324.1 | ||||
| EF426754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607102 | Chr.5:148026584 - 148359140 on Build GRCh38 | Loss |
|
| esv3607106 | Chr.5:148224310 - 148363295 on Build GRCh38 | Loss |
|
| nsv599945 | Chr.5:148269618 - 148340924 on Build GRCh38 | Loss |
|
| dgv726n27 | Chr.5:148268947 - 148310137 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map