Assay Details
Target Gene Details
Entrez Gene ID: | 202559 |
Gene Name: | KH RNA binding domain containing, signal transduction associated 2 |
Gene Aliases: |
SLM-1, SLM1 |
Location: |
Chr.6:61542679-62286227 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHDRBS2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603266 | Chr.6:61471501 - 62189609 on Build GRCh38 | Loss |
|
| nsv603277 | Chr.6:61473996 - 62189609 on Build GRCh38 | Gain |
|
| esv3609206 | Chr.6:61539985 - 61604664 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149764912] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map