Assay Details
Target Gene Details
Entrez Gene ID: | 442247 |
Gene Name: | ret finger protein like 4B |
Gene Aliases: |
RNF211 |
Location: |
Chr.6:112309755-112351480 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFPL4B | XM_017010898.1 | XP_016866387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023049 | Chr.6:112288395 - 113225826 on Build GRCh38 | Gain |
|
| esv3610392 | Chr.6:112315201 - 112337501 on Build GRCh38 | Gain |
|
| nsv1022212 | Chr.6:112222338 - 112365554 on Build GRCh38 | Gain |
|
| esv3571361 | Chr.6:112303348 - 112319912 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map