Assay Details
Target Gene Details
Entrez Gene ID: | 5558 |
Gene Name: | primase (DNA) subunit 2 |
Gene Aliases: |
PRIM2A, p58 |
Location: |
Chr.6:57314805-57646852 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRIM2 | NM_000947.4 | NP_000938.2 | ||
| XM_017011004.1 | XP_016866493.1 | |||
| XM_017011005.1 | XP_016866494.1 | |||
| XM_017011006.1 | XP_016866495.1 | |||
| XM_017011007.1 | XP_016866496.1 | |||
| AK223287.1 | ||||
| AK291915.1 | ||||
| BC064931.1 | AAH64931.1 | |||
| X74331.1 | CAA52378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1184e214 | Chr.6:57341061 - 60368559 on Build GRCh38 | Gain |
|
| esv2759432 | Chr.6:57319743 - 60594857 on Build GRCh38 | Gain |
|
| nsv1077472 | Chr.6:57340901 - 57420102 on Build GRCh38 | Duplication |
|
| dgv10677n54 | Chr.6:57341122 - 60409841 on Build GRCh38 | Gain |
|
| nsv970125 | Chr.6:57341200 - 57420699 on Build GRCh38 | Duplication |
|
| nsv1150365 | Chr.6:57340801 - 57374302 on Build GRCh38 | Duplication |
|
| dgv3320n106 | Chr.6:57340402 - 57432202 on Build GRCh38 | Duplication |
|
| nsv950183 | Chr.6:57341003 - 57432402 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map