Assay Details
Target Gene Details
Entrez Gene ID: | 85236 |
Gene Name: | histone cluster 1, H2bk |
Gene Aliases: |
H2B/S, H2BFAiii, H2BFT, H2BK |
Location: |
Chr.6:27138293-27146858 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2BK | NM_080593.2 | NP_542160.1 | ||
| AJ223352.1 | CAA11276.1 | |||
| BC000893.1 | AAH00893.1 | |||
| BC051872.1 | AAH51872.1 | |||
| BC064959.1 | AAH64959.1 | |||
| BC108737.1 | AAI08738.1 | |||
| DA873786.1 |
More Information
Set Membership: |
|

Genomic Map