Assay Details
Target Gene Details
Entrez Gene ID: | 25998 |
Gene Name: | inhibitor of Bruton tyrosine kinase |
Gene Aliases: |
BTBD26, BTKI |
Location: |
Chr.6:82169987-82248294 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IBTK | NM_001300906.1 | NP_001287835.1 | ||
| NM_015525.3 | NP_056340.2 | |||
| XM_006715453.2 | XP_006715516.1 | |||
| XM_006715454.2 | XP_006715517.1 | |||
| AB037838.2 | ||||
| AK126777.1 | ||||
| AL050018.1 | CAB43239.1 | |||
| BC038244.2 | AAH38244.2 | |||
| BC042171.1 | AAH42171.2 | |||
| BC113696.1 | ||||
| BC113698.1 | ||||
| BC143736.1 | ||||
| BC143745.1 | ||||
| DQ005633.1 | AAY55906.1 | |||
| DQ005634.1 | AAY55907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830713 | Chr.6:82051484 - 82243853 on Build GRCh38 | Gain |
|
| nsv830715 | Chr.6:82096940 - 82246610 on Build GRCh38 | Loss |
|
| nsv7937 | Chr.6:82130518 - 82535071 on Build GRCh38 | Loss |
|
| nsv470837 | Chr.6:82147657 - 82223592 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map