Assay Details
Target Gene Details
Entrez Gene ID: | 90523 |
Gene Name: | muscular LMNA-interacting protein |
Gene Aliases: |
C6orf142 |
Location: |
Chr.6:54018916-54266325 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLIP | NM_001281746.1 | NP_001268675.1 | ||
| NM_138569.2 | NP_612636.2 | |||
| XM_005249476.4 | XP_005249533.1 | |||
| XM_005249478.4 | XP_005249535.1 | |||
| XM_005249480.4 | XP_005249537.1 | |||
| AK055530.1 | BAB70942.1 | |||
| BX647567.1 | ||||
| DA761851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759431 | Chr.6:53974575 - 54142551 on Build GRCh38 | Gain+Loss |
|
| nsv526401 | Chr.6:54031357 - 54047537 on Build GRCh38 | Loss |
|
| nsv1018062 | Chr.6:53977790 - 54060394 on Build GRCh38 | Gain |
|
| nsv603110 | Chr.6:54021120 - 54238996 on Build GRCh38 | Gain |
|
| nsv5307 | Chr.6:54009320 - 54087432 on Build GRCh38 | Deletion |
|
| nsv1034955 | Chr.6:53582809 - 54066257 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75478696] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map