Assay Details
Target Gene Details
Entrez Gene ID: | 221442 |
Gene Name: | adenylate cyclase 10, soluble pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41101034-41140835 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY10P1 | NR_026938.2 | |||
| AK126092.1 | 6 | 3247 | ||
| AK128294.1 | 4 | 1979 | BAC87372.1 | |
| AL833650.1 | ||||
| BX537595.1 | 20 | 5205 | ||
| BX648193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5277 | Chr.6:41112016 - 41146182 on Build GRCh38 | Insertion |
|
| esv3608787 | Chr.6:41134826 - 41139357 on Build GRCh38 | Loss |
|
| nsv602970 | Chr.6:41070039 - 41258521 on Build GRCh38 | Gain |
|
| nsv819921 | Chr.6:41139255 - 41140450 on Build GRCh38 | Loss |
|

Genomic Map