Assay Details
Target Gene Details
Entrez Gene ID: | 116379 |
Gene Name: | interleukin 22 receptor subunit alpha 2 |
Gene Aliases: |
CRF2-10, CRF2-S1, CRF2X, IL-22BP, IL-22R-alpha-2, IL-22RA2, ZCYTOR16 |
Location: |
Chr.6:137143820-137173648 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL22RA2 | NM_052962.2 | NP_443194.1 | ||
| NM_181309.1 | NP_851826.1 | |||
| NM_181310.1 | NP_851827.1 | |||
| AJ297262.1 | CAC83097.1 | |||
| AJ313161.1 | CAC85634.1 | |||
| AJ313162.1 | CAC85635.1 | |||
| AY040566.1 | AAK85714.1 | |||
| AY040567.1 | AAK85715.1 | |||
| AY040568.1 | AAK85716.1 | |||
| AY044429.1 | AAK91775.1 | |||
| AY358111.1 | AAQ88478.1 | |||
| AY358737.1 | AAQ89097.1 | |||
| BC125167.1 | ||||
| BC125168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759473 | Chr.6:137138391 - 137252201 on Build GRCh38 | Loss |
|
| nsv470863 | Chr.6:137080640 - 138014159 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map