Assay Details
Target Gene Details
Entrez Gene ID: | 100287718 |
Gene Name: | ankyrin repeat domain 66 |
Gene Aliases: |
- |
Location: |
Chr.6:46746917-46759218 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD66 | NM_001162435.2 | NP_001155907.2 | ||
| XM_017010148.1 | XP_016865637.1 | |||
| AK057637.1 | ||||
| AK304342.1 | ||||
| BU608132.1 | ||||
| DC417416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428147 | Chr.6:46732445 - 46923866 on Build GRCh38 | Gain |
|

Genomic Map