Assay Details
Target Gene Details
Entrez Gene ID: | 645967 |
Gene Name: | uncharacterized LOC645967 |
Gene Aliases: |
- |
Location: |
Chr.6:149717621-149718896 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC645967 | NR_134652.1 | 1 | 1022 | |
| AB593152.1 | 1 | 1022 | ||
| CA395838.1 | 1 | 278 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732861 | Chr.6:149681836 - 149764933 on Build GRCh38 | Deletion |
|

Genomic Map