Assay Details
Target Gene Details
Entrez Gene ID: | 262 |
Gene Name: | adenosylmethionine decarboxylase 1 |
Gene Aliases: |
ADOMETDC, AMD, SAMDC |
Location: |
Chr.6:110814621-110895713 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMD1 | NM_001287215.1 | NP_001274144.1 | ||
| AK304351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv965752 | Chr.6:110856817 - 110863897 on Build GRCh38 | Duplication |
|
| nsv1032572 | Chr.6:110800096 - 111060820 on Build GRCh38 | Gain |
|
| nsv830773 | Chr.6:110813116 - 110985229 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map