Assay Details
Target Gene Details
Entrez Gene ID: | 23607 |
Gene Name: | CD2 associated protein |
Gene Aliases: |
CMS |
Location: |
Chr.6:47477746-47627263 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD2AP | NM_012120.2 | NP_036252.1 | ||
| XM_005248976.1 | XP_005249033.1 | |||
| XM_017010641.1 | XP_016866130.1 | |||
| AF146277.1 | AAD34595.1 | |||
| BC069444.1 | AAH69444.1 | |||
| DA040834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5290 | Chr.6:47452135 - 47490906 on Build GRCh38 | Insertion |
|
| nsv603012 | Chr.6:47464901 - 47647672 on Build GRCh38 | Loss |
|
| nsv471637 | Chr.6:47339291 - 47493663 on Build GRCh38 | Gain+Loss |
|
| nsv830655 | Chr.6:47407526 - 47595578 on Build GRCh38 | Gain |
|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map