Assay Details
Target Gene Details
Entrez Gene ID: | 347744 |
Gene Name: | chromosome 6 open reading frame 52 |
Gene Aliases: |
- |
Location: |
Chr.6:10671418-10694797 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C6orf52 | NM_001145020.1 | NP_001138492.1 | ||
| NR_026736.1 | ||||
| NR_026737.1 | ||||
| XM_006715088.3 | XP_006715151.1 | |||
| XM_011514572.2 | XP_011512874.1 | |||
| XM_011514574.2 | XP_011512876.1 | |||
| XM_011514575.2 | XP_011512877.1 | |||
| AI376550.1 | ||||
| AI809192.1 | ||||
| AW183566.1 | ||||
| BC016820.1 | AAH16820.1 | |||
| BX280692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv966670 | Chr.6:10447438 - 10871656 on Build GRCh38 | Duplication |
|
| nsv600979 | Chr.6:10449025 - 10839526 on Build GRCh38 | Gain |
|
| nsv470800 | Chr.6:10538711 - 10863092 on Build GRCh38 | Gain |
|
| nsv520853 | Chr.6:10671531 - 10724327 on Build GRCh38 | Loss |
|
| esv3570770 | Chr.6:10685596 - 10689838 on Build GRCh38 | Loss |
|

Genomic Map