Assay Details
Target Gene Details
Entrez Gene ID: | 5167 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 1 |
Gene Aliases: |
ARHR2, COLED, M6S1, NPP1, NPPS, PC-1, PCA1, PDNP1 |
Location: |
Chr.6:131808016-131895155 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP1 | NM_006208.2 | NP_006199.2 | ||
| AK314870.1 | ||||
| BC059375.1 | ||||
| D12485.1 | BAA02054.1 | |||
| M57736.1 | AAA63237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7968 | Chr.6:131596032 - 131999959 on Build GRCh38 | Loss |
|
| esv3610819 | Chr.6:131754426 - 131844936 on Build GRCh38 | Gain |
|
| nsv5479 | Chr.6:131780757 - 131815453 on Build GRCh38 | Deletion |
|
| dgv88n68 | Chr.6:131632788 - 131817256 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map