Assay Details
Target Gene Details
Entrez Gene ID: | 100874282 |
Gene Name: | MLIP intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.6:53998890-54007152 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLIP-IT1 | NR_046832.1 | |||
| CF890740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759431 | Chr.6:53974575 - 54142551 on Build GRCh38 | Gain+Loss |
|
| dgv3316n106 | Chr.6:53990702 - 54001652 on Build GRCh38 | Deletion |
|
| nsv1018062 | Chr.6:53977790 - 54060394 on Build GRCh38 | Gain |
|
| nsv515808 | Chr.6:53990691 - 54000869 on Build GRCh38 | Loss |
|
| esv3609039 | Chr.6:53990701 - 54001629 on Build GRCh38 | Loss |
|
| nsv1034955 | Chr.6:53582809 - 54066257 on Build GRCh38 | Gain |
|

Genomic Map