Assay Details
Target Gene Details
Entrez Gene ID: | 54438 |
Gene Name: | glucose-fructose oxidoreductase domain containing 1 |
Gene Aliases: |
ADG-90, C6orf114 |
Location: |
Chr.6:13357830-13487637 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFOD1 | XM_011514699.2 | 2 | 4434 | XP_011513001.1 |
More Information
Set Membership: |
|

Genomic Map