Assay Details
Target Gene Details
Entrez Gene ID: | 10799 |
Gene Name: | ribonuclease P/MRP subunit p40 |
Gene Aliases: |
RNASEP1, bA428J1.3 |
Location: |
Chr.6:4992405-5004063 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPP40 | NM_001286132.1 | NP_001273061.1 | ||
| NM_001286133.1 | NP_001273062.1 | |||
| NM_006638.3 | NP_006629.2 | |||
| XM_011514253.2 | XP_011512555.1 | |||
| BC017871.1 | AAH17871.1 | |||
| BG430780.1 | ||||
| BM015573.1 | ||||
| BM827175.1 | ||||
| BQ220155.1 | ||||
| BU607286.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600871 | Chr.6:4904146 - 5171775 on Build GRCh38 | Gain |
|
| nsv1033905 | Chr.6:4955592 - 5080670 on Build GRCh38 | Gain |
|
| nsv600872 | Chr.6:4975079 - 5341957 on Build GRCh38 | Gain |
|
| nsv830577 | Chr.6:4834333 - 5050310 on Build GRCh38 | Loss |
|
| esv2422499 | Chr.6:4745194 - 5072988 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map