Assay Details
Target Gene Details
Entrez Gene ID: | 10603 |
Gene Name: | SH2B adaptor protein 2 |
Gene Aliases: |
APS |
Location: |
Chr.7:102284526-102321711 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH2B2 | NM_020979.4 | NP_066189.3 | ||
| XM_005276975.4 | XP_005277032.1 | |||
| XM_005276976.3 | XP_005277033.1 | |||
| XM_005276977.4 | XP_005277034.1 | |||
| XM_011515740.2 | XP_011514042.1 | |||
| XM_017011680.1 | XP_016867169.1 | |||
| AB000520.1 | BAA22514.1 | |||
| BF309652.1 | ||||
| BF434397.1 | ||||
| DN995670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509218 | Chr.7:102146933 - 102338052 on Build GRCh38 | Insertion |
|
| dgv11539n54 | Chr.7:102232361 - 102305844 on Build GRCh38 | Loss |
|
| nsv949918 | Chr.7:102256821 - 102325182 on Build GRCh38 | Deletion |
|
| nsv831082 | Chr.7:102162820 - 102347688 on Build GRCh38 | Gain |
|
| nsv428182 | Chr.7:102257460 - 102735503 on Build GRCh38 | Gain |
|
| dgv6538n100 | Chr.7:102171110 - 103063688 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map