Assay Details
Target Gene Details
Entrez Gene ID: | 221806 |
Gene Name: | von Willebrand factor D and EGF domains |
Gene Aliases: |
- |
Location: |
Chr.7:12330883-12404226 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWDE | NM_001135924.1 | 29 | 5487 | NP_001129396.1 |
| XM_005249641.1 | 26 | 4601 | XP_005249698.1 | |
| XM_011515174.1 | 31 | 5524 | XP_011513476.1 | |
| XM_011515175.1 | 30 | 5386 | XP_011513477.1 | |
| XM_017011828.1 | 27 | 4819 | XP_016867317.1 | |
| XM_017011829.1 | 26 | 4488 | XP_016867318.1 | |
| AK027618.1 | 18 | 3074 | BAB55237.1 | |
| BC144319.1 | 26 | 4640 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6272n100 | Chr.7:12311969 - 12404864 on Build GRCh38 | Loss |
|
| nsv1019021 | Chr.7:12270819 - 12335890 on Build GRCh38 | Loss |
|
| nsv606199 | Chr.7:11466516 - 12447033 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73290465] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map