Assay Details
Target Gene Details
Entrez Gene ID: | 219578 |
Gene Name: | zinc finger protein 804B |
Gene Aliases: |
- |
Location: |
Chr.7:88759368-89337057 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF804B | NM_181646.3 | NP_857597.1 | ||
| AK056672.1 | BAB71248.1 | |||
| AY295769.1 | AAP49447.1 | |||
| BC140826.1 | ||||
| BC140829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6525n100 | Chr.7:88503095 - 90287629 on Build GRCh38 | Gain |
|
| esv3576400 | Chr.7:88517004 - 89889630 on Build GRCh38 | Gain |
|
| esv3576401 | Chr.7:88523463 - 89003741 on Build GRCh38 | Gain |
|
| nsv464626 | Chr.7:88695106 - 90253815 on Build GRCh38 | Gain |
|
| dgv11493n54 | Chr.7:88519150 - 90265948 on Build GRCh38 | Gain |
|
| esv2764019 | Chr.7:88728583 - 89620247 on Build GRCh38 | Loss |
|
| esv2752187 | Chr.7:88546210 - 89018672 on Build GRCh38 | Gain |
|
| dgv6526n100 | Chr.7:88521483 - 89890325 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs370228141] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map