Assay Details
Target Gene Details
Entrez Gene ID: | 8468 |
Gene Name: | FK506 binding protein 6 |
Gene Aliases: |
FKBP36 |
Location: |
Chr.7:73328152-73358637 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP6 | NM_001135211.2 | NP_001128683.1 | ||
| NM_001281304.1 | NP_001268233.1 | |||
| NM_003602.4 | NP_003593.3 | |||
| XM_006716153.2 | XP_006716216.1 | |||
| XM_017012741.1 | XP_016868230.1 | |||
| AF038847.1 | AAC64249.1 | |||
| AF447060.1 | AAP97324.1 | |||
| AK302121.1 | ||||
| BC036817.1 | AAH36817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759537 | Chr.7:72446485 - 73461568 on Build GRCh38 | Gain+Loss |
|
| esv3891145 | Chr.7:73308984 - 73364098 on Build GRCh38 | Gain |
|
| nsv831027 | Chr.7:73173922 - 73442066 on Build GRCh38 | Loss |
|
| dgv6466n100 | Chr.7:73245562 - 73420814 on Build GRCh38 | Gain |
|
| nsv1023935 | Chr.7:73338347 - 73398957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map