Assay Details
Target Gene Details
Entrez Gene ID: | 222236 |
Gene Name: | N-acyl phosphatidylethanolamine phospholipase D |
Gene Aliases: |
FMP30, NAPE-PLD |
Location: |
Chr.7:103099576-103149560 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPEPLD | NM_001122838.1 | NP_001116310.1 | ||
| NM_198990.4 | NP_945341.3 | |||
| XM_005250214.4 | XP_005250271.2 | |||
| XM_005250215.2 | XP_005250272.1 | |||
| XM_005250218.2 | XP_005250275.1 | |||
| XM_011515933.2 | XP_011514235.1 | |||
| XM_011515934.2 | XP_011514236.1 | |||
| XM_017011854.1 | XP_016867343.1 | |||
| XM_017011855.1 | XP_016867344.1 | |||
| XM_017011856.1 | XP_016867345.1 | |||
| XM_017011857.1 | XP_016867346.1 | |||
| AB112352.1 | BAD02399.1 | |||
| AK304644.1 | ||||
| AY357337.1 | AAR13673.1 | |||
| BC071604.1 | AAH71604.1 | |||
| CR936639.1 | CAI56779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752138 | Chr.7:102717873 - 103146688 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map