Assay Details
Target Gene Details
Entrez Gene ID: | 142685 |
Gene Name: | ankyrin repeat and SOCS box containing 15 |
Gene Aliases: |
- |
Location: |
Chr.7:123567003-123637900 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB15 | XM_011515819.2 | XP_011514121.1 | ||
| XM_017011757.1 | XP_016867246.1 |
Target Gene Details
Entrez Gene ID: | 4698 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A5 |
Gene Aliases: |
B13, CI-13KD-B, CI-13kB, NUFM, UQOR13 |
Location: |
Chr.7:123536997-123601651 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA5 | NM_001291304.1 | NP_001278233.1 | ||
| NR_111926.1 | ||||
| BF790429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831116 | Chr.7:123447558 - 123619922 on Build GRCh38 | Gain+Loss |
|
| esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
|
| nsv508479 | Chr.7:123551811 - 123612091 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs184260455] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map