Assay Details
Target Gene Details
Entrez Gene ID: | 9551 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2 |
Gene Aliases: |
ATP5JL |
Location: |
Chr.7:99458161-99466201 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5J2 | NM_001003713.2 | NP_001003713.1 | ||
| NM_001003714.2 | NP_001003714.1 | |||
| NM_001039178.2 | NP_001034267.1 | |||
| NM_004889.3 | NP_004880.1 | |||
| AF047436.1 | AAC39887.1 | |||
| AF088918.1 | AAC34895.1 | |||
| AK223348.1 | BAD97068.1 | |||
| AY046911.1 | AAL06647.1 | |||
| BC003678.1 | AAH03678.1 | |||
| BU587946.1 | ||||
| CB110074.1 | ||||
| CB155273.1 | ||||
| CD173604.1 | ||||
| CD176299.1 | ||||
| CN274665.1 | ||||
| CR456891.1 | CAG33172.1 | |||
| CR542155.1 | CAG46952.1 | |||
| DA064847.1 | ||||
| DB114195.1 | ||||
| EL952520.1 |
Target Gene Details
Entrez Gene ID: | 100526740 |
Gene Name: | ATP5J2-PTCD1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:99416739-99466201 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5J2-PTCD1 | NM_001198879.1 | NP_001185808.1 | ||
| AK295912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73399500] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map