Assay Details
Target Gene Details
Entrez Gene ID: | 84310 |
Gene Name: | chromosome 7 open reading frame 50 |
Gene Aliases: |
YCR016W |
Location: |
Chr.7:978485-1138291 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf50 | NM_001134395.1 | NP_001127867.1 | ||
| NM_001134396.1 | NP_001127868.1 | |||
| NM_001318252.1 | NP_001305181.1 | |||
| NM_032350.5 | NP_115726.1 | |||
| XM_011515581.2 | XP_011513883.1 | |||
| XM_011515582.2 | XP_011513884.1 | |||
| XM_011515583.2 | XP_011513885.1 | |||
| XM_011515584.2 | XP_011513886.1 | |||
| XM_017012720.1 | XP_016868209.1 | |||
| XM_017012721.1 | XP_016868210.1 | |||
| XM_017012722.1 | XP_016868211.1 | |||
| AF495721.1 | AAQ06675.1 | |||
| AW137813.1 | ||||
| BC006224.2 | AAH06224.1 | |||
| BM763235.1 | ||||
| CN261680.1 |
Target Gene Details
Entrez Gene ID: | 115330 |
Gene Name: | G protein-coupled receptor 146 |
Gene Aliases: |
PGR8 |
Location: |
Chr.7:1044573-1059269 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR146 | NM_001303473.1 | NP_001290402.1 | ||
| NM_001303474.1 | NP_001290403.1 | |||
| NM_138445.3 | NP_612454.1 | |||
| XM_005249613.2 | XP_005249670.1 | |||
| XM_006715642.2 | XP_006715705.1 | |||
| XM_006715643.2 | XP_006715706.1 | |||
| AK300166.1 | ||||
| AK313171.1 | ||||
| BI910046.1 | ||||
| CD676083.1 | ||||
| DA169774.1 | ||||
| DB243612.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464245 | Chr.7:886440 - 1065790 on Build GRCh38 | Gain |
|
| nsv605720 | Chr.7:977312 - 1121522 on Build GRCh38 | Loss |
|
| nsv509183 | Chr.7:1040565 - 1097490 on Build GRCh38 | Insertion |
|
| nsv605699 | Chr.7:886312 - 1073232 on Build GRCh38 | Loss |
|
| nsv950784 | Chr.7:855964 - 1074664 on Build GRCh38 | Deletion |
|
| nsv605705 | Chr.7:947983 - 1211575 on Build GRCh38 | Loss |
|
| nsv1161477 | Chr.7:928825 - 1077800 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map