Assay Details
Target Gene Details
Entrez Gene ID: | 389541 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 4 |
Gene Aliases: |
C7orf59 |
Location: |
Chr.7:100130969-100154212 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR4 | NM_001008395.3 | NP_001008396.1 | ||
| NM_001318236.1 | NP_001305165.1 | |||
| NM_001318237.1 | NP_001305166.1 | |||
| NR_134526.1 | ||||
| NR_134527.1 | ||||
| NR_134528.1 | ||||
| NR_134529.1 | ||||
| NR_134530.1 | ||||
| XM_017012198.1 | XP_016867687.1 | |||
| XM_017012199.1 | XP_016867688.1 | |||
| AA419348.1 | ||||
| AA772120.1 | ||||
| AJ714519.1 | ||||
| BC063401.1 | ||||
| BC105609.1 | ||||
| BC130553.1 | ||||
| BC130559.1 | ||||
| BE733059.1 | ||||
| BG706819.1 | ||||
| BG715626.1 | ||||
| BG819624.1 | ||||
| BI598677.1 | ||||
| BI771662.1 | ||||
| CU450474.1 | ||||
| DB498044.1 | ||||
| DB539334.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map