Assay Details
Target Gene Details
Entrez Gene ID: | 8896 |
Gene Name: | BUD31 homolog |
Gene Aliases: |
Cwc14, EDG-2, EDG2, G10, YCR063W, fSAP17 |
Location: |
Chr.7:99408620-99419616 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BUD31 | NM_003910.3 | NP_003901.2 | ||
| XM_005250670.4 | XP_005250727.1 | |||
| XM_005250671.4 | XP_005250728.1 | |||
| XM_005250674.3 | XP_005250731.1 | |||
| XM_017012760.1 | XP_016868249.1 | |||
| XM_017012761.1 | XP_016868250.1 | |||
| XM_017012762.1 | XP_016868251.1 | |||
| AK297784.1 | ||||
| AK316477.1 | ||||
| BC022821.1 | AAH22821.1 | |||
| BC104670.1 | AAI04671.1 | |||
| CR456951.1 | CAG33232.1 | |||
| S77329.1 | AAB33291.1 | |||
| U11861.1 | AAA20008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
|
| nsv5864 | Chr.7:99411680 - 99443573 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map