Assay Details
Target Gene Details
Entrez Gene ID: | 389541 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 4 |
Gene Aliases: |
C7orf59 |
Location: |
Chr.7:100130969-100154212 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR4 | XM_017012198.1 | XP_016867687.1 | ||
| XM_017012199.1 | XP_016867688.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv966865 | Chr.7:100137962 - 100142855 on Build GRCh38 | Duplication |
|
| esv3614290 | Chr.7:100138319 - 100141368 on Build GRCh38 | Loss |
|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map