Assay Details
Target Gene Details
Entrez Gene ID: | 5764 |
Gene Name: | pleiotrophin |
Gene Aliases: |
HARP, HB-GAM, HBBM, HBGF-8, HBGF8, HBNF, HBNF-1, NEGF1, OSF-1 |
Location: |
Chr.7:137227341-137343865 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTN | NM_001321386.1 | NP_001308315.1 | ||
| NM_001321387.1 | NP_001308316.1 | |||
| NM_002825.5 | NP_002816.1 | |||
| XM_017012473.1 | XP_016867962.1 | |||
| AK290488.1 | ||||
| AK313424.1 | ||||
| BC005916.1 | AAH05916.1 | |||
| BI462078.1 | ||||
| BT019692.1 | AAV38498.1 | |||
| BX441769.2 | ||||
| CN413530.1 | ||||
| CR450338.1 | CAG29334.1 | |||
| D90226.1 | BAA14261.1 | |||
| M57399.1 | AAA35961.1 | |||
| X52946.1 | CAA37121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv11620n54 | Chr.7:137198178 - 137250712 on Build GRCh38 | Loss |
|
| esv33947 | Chr.7:137235266 - 138283104 on Build GRCh38 | Gain |
|
| nsv831149 | Chr.7:137146918 - 137321721 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75746312] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map