Assay Details
Target Gene Details
Entrez Gene ID: | 100526740 |
Gene Name: | ATP5J2-PTCD1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:99416739-99466201 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5J2-PTCD1 | NM_001198879.1 | NP_001185808.1 | ||
| AK295912.1 |
Target Gene Details
Entrez Gene ID: | 10898 |
Gene Name: | cleavage and polyadenylation specific factor 4 |
Gene Aliases: |
CPSF30, NAR, NEB-1, NEB1 |
Location: |
Chr.7:99438920-99457377 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPSF4 | NM_001081559.2 | NP_001075028.1 | ||
| NM_001318160.1 | NP_001305089.1 | |||
| NM_001318161.1 | NP_001305090.1 | |||
| NM_001318162.1 | NP_001305091.1 | |||
| NM_006693.3 | NP_006684.1 | |||
| XM_011515755.2 | XP_011514057.1 | |||
| XM_011515756.2 | XP_011514058.1 | |||
| XM_011515757.2 | XP_011514059.1 | |||
| XM_017011700.1 | XP_016867189.1 | |||
| XM_017011701.1 | XP_016867190.1 | |||
| XM_017011702.1 | XP_016867191.1 | |||
| AK301745.1 | ||||
| BC003101.1 | AAH03101.1 | |||
| BC050738.1 | AAH50738.1 | |||
| BM757197.1 | ||||
| BQ052009.1 | ||||
| BX358595.2 | ||||
| CR542161.1 | CAG46958.1 | |||
| CX754488.1 | ||||
| U79569.1 | AAD00321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
|
| nsv5864 | Chr.7:99411680 - 99443573 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map