Assay Details
Target Gene Details
Entrez Gene ID: | 7551 |
Gene Name: | zinc finger protein 3 |
Gene Aliases: |
A8-51, HF.12, KOX25, PP838, Zfp113 |
Location: |
Chr.7:100063847-100082548 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF3 | NM_001278284.1 | NP_001265213.1 | ||
| NM_001278287.1 | NP_001265216.1 | |||
| NM_001278290.1 | NP_001265219.1 | |||
| NM_001278291.1 | NP_001265220.1 | |||
| NM_001278292.1 | NP_001265221.1 | |||
| NM_001318135.1 | NP_001305064.1 | |||
| NM_001318136.1 | NP_001305065.1 | |||
| NM_001318137.1 | NP_001305066.1 | |||
| NM_017715.3 | NP_060185.2 | |||
| NM_032924.4 | NP_116313.3 | |||
| XM_006716111.1 | XP_006716174.1 | |||
| XM_017012582.1 | XP_016868071.1 | |||
| AF193057.1 | AAG22485.1 | |||
| AF217988.1 | AAG17231.1 | |||
| AK091984.1 | ||||
| AK297625.1 | ||||
| AK309719.1 | ||||
| AK315479.1 | ||||
| BC011887.2 | AAH11887.2 | |||
| BC013603.1 | ||||
| BQ071498.1 | ||||
| CB129643.1 | ||||
| CB988353.1 | ||||
| DA443888.1 | ||||
| DA623743.1 | ||||
| DC347416.1 | ||||
| DC384519.1 | ||||
| HY096067.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map