Assay Details
Target Gene Details
Entrez Gene ID: | 10434 |
Gene Name: | lysophospholipase I |
Gene Aliases: |
APT-1, APT1, LPL-I, LPL1, hAPT1 |
Location: |
Chr.8:54046367-54102017 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYPLA1 | NM_001279356.1 | NP_001266285.1 | ||
| NM_001279357.1 | NP_001266286.1 | |||
| NM_001279358.1 | NP_001266287.1 | |||
| NM_001279359.1 | NP_001266288.1 | |||
| NM_001279360.1 | NP_001266289.1 | |||
| NM_006330.3 | NP_006321.1 | |||
| XM_005251127.4 | XP_005251184.1 | |||
| XM_017012956.1 | XP_016868445.1 | |||
| XM_017012957.1 | XP_016868446.1 | |||
| XM_017012958.1 | XP_016868447.1 | |||
| XM_017012959.1 | XP_016868448.1 | |||
| XM_017012960.1 | XP_016868449.1 | |||
| AA307642.1 | ||||
| AF052112.1 | ||||
| AF077198.1 | AAD26993.1 | |||
| AF077199.1 | AAD26994.1 | |||
| AF081281.1 | AAC31610.1 | |||
| AF291053.1 | AAG10063.1 | |||
| AK289400.1 | ||||
| AK298206.1 | ||||
| AK313739.1 | ||||
| BC008652.2 | AAH08652.1 | |||
| BC010397.1 | AAH10397.1 | |||
| CR457103.1 | CAG33384.1 | |||
| DA817192.1 | ||||
| DA881616.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759617 | Chr.8:53920430 - 54104405 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map