Assay Details
Target Gene Details
Entrez Gene ID: | 729330 |
Gene Name: | otoconin 90 |
Gene Aliases: |
PLA2L |
Location: |
Chr.8:132024220-132059380 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OC90 | NM_001080399.2 | NP_001073868.2 | ||
| AK297911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018172 | Chr.8:131797977 - 132121114 on Build GRCh38 | Gain |
|
| nsv612325 | Chr.8:131918447 - 132338873 on Build GRCh38 | Gain |
|
| esv3618853 | Chr.8:131375690 - 132136917 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map